The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029210.1 | NP_083486.1 | 3 - 4 | 993 | 65 |
GenBank mRNA | AK173092.1 | - | 3 - 4 | 993 | 65 |
BC132111.1 | - | 2 - 3 | 829 | 65 | |
BC137861.1 | - | 2 - 3 | 829 | 65 |