Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptcd2
Entrez Gene ID
68927
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
1190005P08Rik
Chromosome Location
Chr.13: 99319649 - 99344678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026873.2 NP_081149.1 6 - 7 623 114
GenBank mRNA AK004502.1 - 6 - 7 633 113
AK144396.1 - 6 - 7 635 114
BC016563.1 - 6 - 7 625 114
BC025110.1 - 6 - 7 623 114

View Full Product Details