Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rad50
Entrez Gene ID
19360
Gene Name
RAD50 homolog (S. cerevisiae)
Gene Aliases
Mrell, Rad50l
Chromosome Location
Chr.11: 53649519 - 53707319 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009012.2 NP_033038.2 19 - 20 3305 126

View Full Product Details