Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phc3
Entrez Gene ID
241915
Gene Name
polyhomeotic-like 3 (Drosophila)
Gene Aliases
E030046K01Rik, Edr3, Hph3
Chromosome Location
Chr.3: 30899295 - 30969479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165954.1 NP_001159426.1 4 - 5 441 94
NM_001165955.1 NP_001159427.1 4 - 5 456 94
NM_001165956.1 NP_001159428.1 4 - 5 441 94
NM_153421.2 NP_700470.2 4 - 5 456 94
GenBank mRNA AJ414610.1 - 4 - 5 442 94
AK037348.1 - 4 - 5 432 94
AK087339.1 - 4 - 5 447 94
BC132533.1 - 4 - 5 425 94
BC145123.1 - 4 - 5 425 94
BC145124.1 - 4 - 5 425 94

View Full Product Details