Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Phc3
Entrez Gene ID
241915
Gene Name
polyhomeotic-like 3 (Drosophila)
Gene Aliases
E030046K01Rik, Edr3, Hph3
Chromosome Location
Chr.3: 30899295 - 30969479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001165954.1 NP_001159426.1 10 - 11 2219 96
NM_001165955.1 NP_001159427.1 9 - 10 2135 96
NM_001165956.1 NP_001159428.1 11 - 12 2093 96
NM_153421.2 NP_700470.2 10 - 11 2234 96
GenBank mRNA AJ414610.1 - 10 - 11 2220 96
AK037348.1 - 11 - 12 2279 96
AK087339.1 - 9 - 10 2129 96
AK166208.1 - 10 - 11 2014 96
BC044839.1 - 3 - 4 920 96
BC132533.1 - 10 - 11 2203 96
BC145123.1 - 11 - 12 2077 96
BC145124.1 - 11 - 12 2077 96

View Full Product Details