Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Intu
Entrez Gene ID
380614
Gene Name
inturned planar cell polarity effector homolog (Drosophila)
Gene Aliases
9230116I04Rik, 9430087H23Rik, Pdzd6, Pdzk6, mKIAA1284
Chromosome Location
Chr.3: 40653702 - 40704774 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175515.5 NP_780724.3 - 1553 63
GenBank mRNA AK035092.1 - - 1534 63
AK122485.1 - 7 - 8 1093 63
BC125523.1 - - 1500 63
BC132387.1 - - 1500 63

View Full Product Details