The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178038.1 | NP_001171509.1 | 4 - 5 | 720 | 67 |
GenBank mRNA | BC051402.1 | - | 5 - 6 | 807 | 67 |