The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122675.1 | NP_001116147.1 | 5 - 6 | 1682 | 107 |
NM_001122676.1 | NP_001116148.1 | 5 - 6 | 1682 | 107 | |
GenBank mRNA | AK046562.1 | - | 5 - 6 | 1683 | 107 |
AK173248.1 | - | 5 - 6 | 1399 | 107 | |
BC035528.1 | - | 2 - 3 | 172 | 107 | |
BC082306.1 | - | 2 - 3 | 174 | 107 |