The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001122675.1 | NP_001116147.1 | 7 - 8 | 1845 | 138 |
NM_001122676.1 | NP_001116148.1 | 7 - 8 | 1845 | 138 | |
GenBank mRNA | AK046562.1 | - | 7 - 8 | 1846 | 138 |
AK173248.1 | - | 7 - 8 | 1562 | 138 | |
BC035528.1 | - | 4 - 5 | 335 | 138 |