Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Lemd1
- Entrez Gene ID
-
213409
- Gene Name
- LEM domain containing 1
- Gene Aliases
-
4930540I23Rik, 6430514M23Rik, AI428128, LEMP-1
- Chromosome Location
-
Chr.1: 132190687 - 132257382 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK144269.1 |
- |
- |
305 |
94 |
View Full Product Details