Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srgap2
Entrez Gene ID
14270
Gene Name
SLIT-ROBO Rho GTPase activating protein 2
Gene Aliases
9930124L22Rik, AI448945, FBP2, Fnbp2, srGAP3
Chromosome Location
Chr.1: 131285251 - 131527369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081011.2 NP_001074480.2 19 - 20 2902 60
GenBank mRNA AK132220.1 - 5 - 6 575 60
AK161932.1 - 8 - 9 806 60
BC030457.1 - 18 - 19 2209 60
BC151081.1 - 18 - 19 2520 60
BC151082.1 - 18 - 19 2520 60
BC158055.1 - 18 - 19 2518 60
BC172152.1 - 18 - 19 2520 60

View Full Product Details