Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cntnap5a
Entrez Gene ID
636808
Gene Name
contactin associated protein-like 5A
Gene Aliases
Caspr5-1, EG636808
Chromosome Location
Chr.1: 115684497 - 116587323 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077425.1 NP_001070893.1 14 - 15 2230 83

View Full Product Details