Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ano7
Entrez Gene ID
404545
Gene Name
anoctamin 7
Gene Aliases
Ngep, Ngep-L, Pcanap5, Tmem16g
Chromosome Location
Chr.1: 93373796 - 93404304 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271884.1 NP_001258813.1 2 - 3 270 134
NM_207031.2 NP_996914.1 2 - 3 270 134
GenBank mRNA BC116706.1 - 2 - 3 236 134

View Full Product Details