The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028568.1 | NP_082844.1 | 1 - 1 | 743 | 85 |
GenBank mRNA | AK007061.1 | - | 1 - 1 | 756 | 85 |
BC048605.1 | - | 1 - 1 | 743 | 85 |