Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Steap3
Entrez Gene ID
68428
Gene Name
STEAP family member 3
Gene Aliases
1010001D01Rik, Tsap6, pHyde
Chromosome Location
Chr.1: 120226414 - 120271082 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK171237.1 - 4 - 5 1249 100

View Full Product Details