Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdkl3
Entrez Gene ID
213084
Gene Name
cyclin-dependent kinase-like 3
Gene Aliases
AI852479, B230379H01Rik
Chromosome Location
Chr.11: 52000858 - 52089778 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BB759970.1 - 3 - 4 285 117

View Full Product Details