The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166653.1 | NP_001160125.1 | 9 - 10 | 1580 | 123 |
NM_001166654.1 | NP_001160126.1 | 9 - 10 | 1670 | 123 | |
NM_001166656.1 | NP_001160128.1 | 9 - 10 | 1580 | 123 | |
NM_153785.4 | NP_722480.3 | 9 - 10 | 1670 | 123 | |
GenBank mRNA | AK043995.1 | - | 1 - 2 | 264 | 123 |
AK045036.1 | - | 8 - 9 | 2028 | 123 | |
AK045356.1 | - | 9 - 10 | 1570 | 123 | |
AK046394.1 | - | 9 - 10 | 1671 | 123 | |
AK078804.1 | - | 9 - 10 | 1562 | 123 |