Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pik3ca
Entrez Gene ID
18706
Gene Name
phosphatidylinositol 3-kinase, catalytic, alpha polypeptide
Gene Aliases
6330412C24Rik, caPI3K, p110, p110alpha
Chromosome Location
Chr.3: 32397055 - 32468486 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008839.2 NP_032865.2 5 - 6 1200 107
GenBank mRNA AK051885.1 - 5 - 6 833 107
BC089038.1 - 5 - 6 1200 107
BC130228.1 - 5 - 6 1149 107

View Full Product Details