Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccna1
Entrez Gene ID
12427
Gene Name
cyclin A1
Gene Aliases
-
Chromosome Location
Chr.3: 55045469 - 55055330 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007628.3 NP_031654.2 2 - 3 277 114
GenBank mRNA AK077114.1 - 2 - 3 283 114
BC120518.1 - 1 - 2 192 114
BC125436.1 - 1 - 2 192 114
X84311.1 - 2 - 3 276 114

View Full Product Details