Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccna1
Entrez Gene ID
12427
Gene Name
cyclin A1
Gene Aliases
-
Chromosome Location
Chr.3: 55045469 - 55055330 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007628.3 NP_031654.2 6 - 7 1077 63
GenBank mRNA AK077114.1 - 6 - 7 1083 63
BC120518.1 - 5 - 6 992 63
BC125436.1 - 5 - 6 992 63
X84311.1 - 6 - 7 1076 63

View Full Product Details