The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081295.1 | NP_001074764.1 | 1 - 2 | 1792 | 82 |
GenBank mRNA | AK144976.1 | - | 1 - 2 | 1298 | 82 |