Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fstl4
Entrez Gene ID
320027
Gene Name
follistatin-like 4
Gene Aliases
B230374F23Rik, SPIG1, mKIAA1061
Chromosome Location
Chr.11: 52764044 - 53192450 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177059.3 NP_796033.2 12 - 13 1641 78
GenBank mRNA AF374459.1 - 12 - 13 1641 78
AK046350.1 - 12 - 13 1692 78
AK081114.1 - 12 - 13 1691 78
AK220367.1 - 8 - 9 1021 78
BC132353.1 - 12 - 13 1618 78
BC144824.1 - 12 - 13 1611 78

View Full Product Details