Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Msh3
Entrez Gene ID
17686
Gene Name
mutS homolog 3 (E. coli)
Gene Aliases
D13Em1, Rep-3, Rep3
Chromosome Location
Chr.13: 92211879 - 92355003 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010829.2 NP_034959.2 22 - 23 3108 81
GenBank mRNA AK133907.1 - 22 - 23 3100 81
AK161089.1 - 22 - 23 3104 81
AK161430.1 - 4 - 5 526 81
BC025063.1 - 4 - 5 723 81
BC040784.1 - 7 - 8 959 81
BC150759.1 - 22 - 23 3027 81
CK022909.1 - - 582 81
M80360.1 - 22 - 23 3093 81

View Full Product Details