The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010558.1 | NP_034688.1 | 3 - 4 | 351 | 133 |
GenBank mRNA | BC125366.1 | - | 3 - 4 | 336 | 133 |
BC132066.1 | - | 3 - 4 | 336 | 133 | |
X04601.1 | - | 3 - 4 | 351 | 133 | |
X06270.1 | - | 3 - 4 | 351 | 133 |