Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kdsr
Entrez Gene ID
70750
Gene Name
3-ketodihydrosphingosine reductase
Gene Aliases
6330410P18Rik, 9430079B08Rik, Fvt1
Chromosome Location
Chr.1: 106720410 - 106759830 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027534.2 NP_081810.1 5 - 6 511 75
GenBank mRNA AK018155.1 - 6 - 7 486 75
AK170064.1 - 5 - 6 394 75
AY634684.1 - 5 - 6 420 75
BC023820.1 - 5 - 6 501 75

View Full Product Details