Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pm20d1
Entrez Gene ID
212933
Gene Name
peptidase M20 domain containing 1
Gene Aliases
4732466D17Rik, AI098026
Chromosome Location
Chr.1: 131797385 - 131821419 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178079.3 NP_835180.2 3 - 4 512 62
GenBank mRNA AK028883.1 - 3 - 4 509 62
AK128969.1 - 3 - 4 518 62
AK143953.1 - 3 - 4 507 62
BC120725.1 - 3 - 4 512 62
BC125395.1 - 3 - 4 512 62

View Full Product Details