Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Atp1a2
Entrez Gene ID
98660
Gene Name
ATPase, Na+/K+ transporting, alpha 2 polypeptide
Gene Aliases
AW060654, Atpa-3, mKIAA0778
Chromosome Location
Chr.1: 172271709 - 172298064 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178405.3 NP_848492.1 22 - 23 3207 99
GenBank mRNA AK137602.1 - 22 - 23 3143 99
AK147331.1 - 22 - 23 3139 99
AK148775.1 - 21 - 22 3329 99
AK161840.1 - 22 - 23 3139 99
AK164509.1 - 22 - 23 3143 99
AK168703.1 - 22 - 23 3139 99
BC025807.1 - 11 - 12 1435 99
BC036127.1 - 22 - 23 3131 99
BC041774.1 - 19 - 20 2857 99

View Full Product Details