Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gorab
Entrez Gene ID
98376
Gene Name
golgin, RAB6-interacting
Gene Aliases
AI467484, NTKL-BP1, Scyl1bp1
Chromosome Location
Chr.1: 163384906 - 163403908 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK039170.1 - 2 - 3 487 69
AK170455.1 - 2 - 3 497 69

View Full Product Details