Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fmo6
Entrez Gene ID
226565
Gene Name
flavin containing monooxygenase 6
Gene Aliases
-
Chromosome Location
Chr.1: 162916551 - 162937572 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC051402.1 - 3 - 4 439 81

View Full Product Details