Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dusp27
Entrez Gene ID
240892
Gene Name
dual specificity phosphatase 27 (putative)
Gene Aliases
C130085G02Rik, Gm209
Chromosome Location
Chr.1: 166098148 - 166127898 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033344.3 NP_001028516.2 1 - 2 105 105
GenBank mRNA AK142263.1 - 1 - 2 106 105
BY730343.1 - 1 - 2 106 105

View Full Product Details