The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011495.2 | NP_035625.2 | 1 - 2 | 325 | 75 |
NM_173169.2 | NP_775261.2 | 1 - 2 | 325 | 75 | |
NR_045576.1 | 1 - 2 | 325 | 75 | ||
GenBank mRNA | AK006827.1 | - | 1 - 2 | 231 | 75 |
AK045082.1 | - | 1 - 2 | 249 | 75 | |
AK137080.1 | - | 1 - 2 | 237 | 75 | |
AK137471.1 | - | 1 - 2 | 236 | 75 | |
AK164465.1 | - | 1 - 2 | 235 | 75 | |
BC026785.1 | - | 1 - 2 | 235 | 75 | |
BC057940.1 | - | 1 - 2 | 222 | 75 | |
L29479.1 | - | 1 - 2 | 227 | 75 | |
L29480.1 | - | 1 - 2 | 227 | 75 |