Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fat4
Entrez Gene ID
329628
Gene Name
FAT tumor suppressor homolog 4 (Drosophila)
Gene Aliases
6030410K14Rik, 9430004M15
Chromosome Location
Chr.3: 38886487 - 39011987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183221.3 NP_899044.3 7 - 8 7210 66
GenBank mRNA DQ286572.1 - 7 - 8 7210 66

View Full Product Details