Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lhx9
Entrez Gene ID
16876
Gene Name
LIM homeobox protein 9
Gene Aliases
3110009O07Rik, BB104635, LH2B
Chromosome Location
Chr.1: 138825186 - 138848649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025565.2 NP_001020736.1 1 - 2 638 72
NM_001042577.1 NP_001036042.1 1 - 2 638 72
NM_010714.3 NP_034844.1 3 - 4 265 72
GenBank mRNA AB221560.1 - 2 - 3 148 72
AF134761.1 - 3 - 4 264 72
AJ243851.1 - 3 - 4 264 72
AK012930.1 - 1 - 2 76 72
AK031478.1 - 1 - 2 640 72
BC072623.1 - 1 - 2 651 72

View Full Product Details