Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Exosc8
Entrez Gene ID
69639
Gene Name
exosome component 8
Gene Aliases
2310032N20Rik, CIP3, mKIAA4013
Chromosome Location
Chr.3: 54728679 - 54735364 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163570.1 NP_001157042.1 9 - 10 959 113
NM_027148.3 NP_081424.3 9 - 10 947 113
GenBank mRNA AJ505005.1 - 9 - 10 607 113
AK009584.1 - 9 - 10 619 113
AK169719.1 - 7 - 8 550 113
AK220227.1 - 9 - 10 947 113
BC004769.1 - 6 - 7 1996 113
BC067250.1 - 8 - 9 590 113

View Full Product Details