The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001168321.1 | NP_001161793.1 | 13 - 14 | 4941 | 79 |
NM_029510.3 | NP_083786.2 | 14 - 15 | 5766 | 79 | |
NM_175044.3 | NP_778209.2 | 13 - 14 | 5712 | 79 | |
NM_175045.3 | NP_778210.2 | 14 - 15 | 5664 | 79 | |
NM_175046.3 | NP_778211.2 | 13 - 14 | 5610 | 79 | |
GenBank mRNA | AK018034.1 | - | 4 - 5 | 531 | 79 |
AK031282.1 | - | 6 - 7 | 1068 | 79 | |
AK129398.1 | - | 13 - 14 | 4941 | 79 | |
AY161170.1 | - | 14 - 15 | 5081 | 79 | |
AY161171.1 | - | 13 - 14 | 5027 | 79 | |
AY161172.1 | - | 14 - 15 | 4979 | 79 | |
AY161173.1 | - | 13 - 14 | 4925 | 79 | |
BC038637.1 | - | 6 - 7 | 971 | 79 | |
BC058656.1 | - | 14 - 15 | 5165 | 79 |