Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcor
Entrez Gene ID
71458
Gene Name
BCL6 interacting corepressor
Gene Aliases
5830466J11Rik, 8430401K06Rik, BcorR, D930024N20Rik, mKIAA1575
Chromosome Location
Chr.X: 12036737 - 12160355 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168321.1 NP_001161793.1 3 - 4 275 78
NM_029510.3 NP_083786.2 3 - 4 944 78
NM_175044.3 NP_778209.2 3 - 4 944 78
NM_175045.3 NP_778210.2 3 - 4 944 78
NM_175046.3 NP_778211.2 3 - 4 944 78
GenBank mRNA AK053309.1 - 3 - 4 945 78
AK086385.2 - 3 - 4 451 78
AK129398.1 - 3 - 4 275 78
AY161170.1 - 3 - 4 259 78
AY161171.1 - 3 - 4 259 78
AY161172.1 - 3 - 4 259 78
AY161173.1 - 3 - 4 259 78
BC058656.1 - 3 - 4 445 78

View Full Product Details