Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ifi47
Entrez Gene ID
15953
Gene Name
interferon gamma inducible protein 47
Gene Aliases
47kDa, IRG-47, Ifggc1, Igrd, Iigp4, Iipg4, Irgd
Chromosome Location
Chr.11: 49076587 - 49096987 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271676.1 NP_001258605.1 2 - 3 343 107

View Full Product Details