Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Zc3h11a
- Entrez Gene ID
-
70579
- Gene Name
- zinc finger CCCH type containing 11A
- Gene Aliases
-
1110003F06Rik, 5730454B08Rik, G630041M05Rik, Zc3hdc11a, Zc3hh11a, mKIAA0663
- Chromosome Location
-
Chr.1: 133619868 - 133661399 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK017663.1 |
- |
2 - 3 |
181 |
114 |
View Full Product Details