The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153114.2 | NP_694754.1 | 3 - 4 | 377 | 61 |
GenBank mRNA | AK083090.1 | - | 2 - 3 | 2665 | 61 |
AY078071.1 | - | 2 - 3 | 125 | 61 | |
BC100323.1 | - | 3 - 4 | 377 | 61 | |
BC132494.1 | - | 2 - 3 | 296 | 61 | |
BC132498.1 | - | 2 - 3 | 296 | 61 |