The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001160018.1 | NP_001153490.1 | 1 - 2 | 717 | 72 |
NM_144791.2 | NP_659040.2 | 1 - 2 | 717 | 72 | |
GenBank mRNA | AB251963.1 | - | 1 - 2 | 657 | 72 |
AK139709.1 | - | 1 - 2 | 386 | 72 | |
AK149727.1 | - | 1 - 2 | 637 | 72 | |
AK152751.1 | - | 1 - 2 | 151 | 72 | |
BC010841.1 | - | 1 - 2 | 532 | 72 | |
BQ891636.1 | - | 1 - 2 | 312 | 72 |