Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sohlh2
Entrez Gene ID
74434
Gene Name
spermatogenesis and oogenesis specific basic helix-loop-helix 2
Gene Aliases
4933406N12Rik, AI413378
Chromosome Location
Chr.3: 55182044 - 55209957 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028937.3 NP_083213.2 3 - 4 349 89
GenBank mRNA AK016704.1 - 3 - 4 351 89
BC120917.1 - 3 - 4 344 89
BC120918.2 - 3 - 4 344 89
DQ086115.1 - 3 - 4 349 89

View Full Product Details