The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045530.2 | NP_001038995.1 | 3 - 4 | 533 | 74 |
GenBank mRNA | AK143205.1 | - | 3 - 4 | 499 | 74 |
BC139344.1 | - | 2 - 3 | 294 | 74 | |
BC139345.1 | - | 2 - 3 | 294 | 74 |