The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010907.2 | NP_035037.2 | 5 - 6 | 1016 | 87 |
GenBank mRNA | AK151115.1 | - | 5 - 6 | 969 | 87 |
AK151608.1 | - | 5 - 6 | 969 | 87 | |
AK161622.1 | - | 5 - 6 | 928 | 87 | |
BC046754.1 | - | 5 - 6 | 949 | 87 | |
U36277.1 | - | 5 - 6 | 960 | 87 |