The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013626.3 | NP_038654.2 | 21 - 22 | 2789 | 109 |
GenBank mRNA | AK132581.1 | - | 9 - 10 | 1215 | 109 |
BC129869.1 | - | 4 - 5 | 469 | 109 | |
U79523.1 | - | 21 - 22 | 2674 | 109 |