Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mccc1
Entrez Gene ID
72039
Gene Name
methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases
1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106
Chromosome Location
Chr.3: 35959296 - 36000678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023644.4 NP_076133.3 17 - 18 2091 73
GenBank mRNA AF310338.1 - 17 - 18 2012 73
AK007782.1 - 17 - 18 2044 73
AK031072.1 - 17 - 18 2052 73
AK032203.1 - 17 - 18 2050 73
AK168589.1 - 17 - 18 1980 73
BC021382.1 - 17 - 18 2055 73

View Full Product Details