Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mccc1
Entrez Gene ID
72039
Gene Name
methylcrotonoyl-Coenzyme A carboxylase 1 (alpha)
Gene Aliases
1810045E08Rik, 2310058B18Rik, MCCalpha, Mcca, R75106
Chromosome Location
Chr.3: 35959296 - 36000678 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023644.4 NP_076133.3 18 - 19 2156 70
GenBank mRNA AF310338.1 - 18 - 19 2077 70
AK007782.1 - 18 - 19 2109 70
AK031072.1 - 18 - 19 2117 70
AK032203.1 - 18 - 19 2115 70
AK168589.1 - 18 - 19 2045 70
BC021382.1 - 18 - 19 2120 70

View Full Product Details