The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011063.2 | NP_035193.1 | 2 - 3 | 359 | 78 |
GenBank mRNA | AK077421.1 | - | 2 - 3 | 288 | 78 |
AK089070.1 | - | 2 - 3 | 287 | 78 | |
AK153141.1 | - | 2 - 3 | 292 | 78 | |
AK153493.1 | - | 2 - 3 | 290 | 78 | |
AK161635.1 | - | 2 - 3 | 275 | 78 | |
AK161977.1 | - | 2 - 3 | 288 | 78 | |
BC038282.1 | - | 2 - 3 | 267 | 78 | |
G49234.1 | - | 2 - 3 | 281 | 78 |