Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptprv
Entrez Gene ID
13924
Gene Name
protein tyrosine phosphatase, receptor type, V
Gene Aliases
Esp, OST, OST-PTP, mOST-PTP
Chromosome Location
Chr.1: 135108498 - 135132575 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007955.3 NP_031981.3 30 - 31 5187 73
GenBank mRNA AF300701.1 - 29 - 30 4664 73
AK156112.1 - 30 - 31 5190 73
U36488.1 - 26 - 27 4349 73

View Full Product Details