Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ptprv
Entrez Gene ID
13924
Gene Name
protein tyrosine phosphatase, receptor type, V
Gene Aliases
Esp, OST, OST-PTP, mOST-PTP
Chromosome Location
Chr.1: 135108498 - 135132575 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007955.3 NP_031981.3 4 - 5 828 89
GenBank mRNA AK156112.1 - 4 - 5 830 89

View Full Product Details