Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mettl11b
Entrez Gene ID
240879
Gene Name
methyltransferase like 11B
Gene Aliases
Gm207, NTM1B
Chromosome Location
Chr.1: 163701870 - 163725232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001143956.1 NP_001137428.1 1 - 2 334 74
GenBank mRNA BC157906.1 - 1 - 2 334 74
BC158115.1 - 1 - 2 311 74
BC172048.1 - 1 - 2 334 74

View Full Product Details